missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 2-Fluorobenzoate 98.0+%, TCI America™
$55.92 - $544.30
Chemical Identifiers
| CAS | 394-35-4 |
|---|---|
| Molecular Formula | C8H7FO2 |
| Molecular Weight (g/mol) | 154.14 |
| MDL Number | MFCD00017913 |
| InChI Key | QAFJIJWLEBLXHH-UHFFFAOYSA-N |
| Synonym | methyl o-fluorobenzoate, methyl-2-fluorobenzoate, 2-fluorobenzoic acid methyl ester, benzoic acid, 2-fluoro-, methyl ester, o-fluorobenzoic acid, methyl ester, benzoic acid, o-fluoro-, methyl ester, 2-fluoro-benzoic acid methyl ester, methyl2-fluorobenzoate, pubchem3565, acmc-209j5e |
| PubChem CID | 67854 |
| IUPAC Name | methyl 2-fluorobenzoate |
| SMILES | COC(=O)C1=CC=CC=C1F |
Chemical Identifiers
| 394-35-4 | |
| 154.14 | |
| QAFJIJWLEBLXHH-UHFFFAOYSA-N | |
| 67854 | |
| COC(=O)C1=CC=CC=C1F |
| C8H7FO2 | |
| MFCD00017913 | |
| methyl o-fluorobenzoate, methyl-2-fluorobenzoate, 2-fluorobenzoic acid methyl ester, benzoic acid, 2-fluoro-, methyl ester, o-fluorobenzoic acid, methyl ester, benzoic acid, o-fluoro-, methyl ester, 2-fluoro-benzoic acid methyl ester, methyl2-fluorobenzoate, pubchem3565, acmc-209j5e | |
| methyl 2-fluorobenzoate |
Specifications
| 394-35-4 | |
| 102°C | |
| MFCD00017913 | |
| methyl o-fluorobenzoate, methyl-2-fluorobenzoate, 2-fluorobenzoic acid methyl ester, benzoic acid, 2-fluoro-, methyl ester, o-fluorobenzoic acid, methyl ester, benzoic acid, o-fluoro-, methyl ester, 2-fluoro-benzoic acid methyl ester, methyl2-fluorobenzoate, pubchem3565, acmc-209j5e | |
| COC(=O)C1=CC=CC=C1F | |
| 154.14 | |
| 154.14 | |
| Liquid |
| Colorless | |
| C8H7FO2 | |
| 25 g | |
| QAFJIJWLEBLXHH-UHFFFAOYSA-N | |
| methyl 2-fluorobenzoate | |
| 67854 | |
| ≥98.0% (GC) | |
| Methyl 2-Fluorobenzoate |
Safety and Handling
TSCA : Yes