missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-Fluorobenzoate 98.0+%, TCI America™

Catalog No. F024125G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F024125G 25 g
F0241500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F024125G Supplier TCI America Supplier No. F024125G
Only null left

Chemical Identifiers

CAS 394-35-4
Molecular Formula C8H7FO2
Molecular Weight (g/mol) 154.14
MDL Number MFCD00017913
InChI Key QAFJIJWLEBLXHH-UHFFFAOYSA-N
Synonym methyl o-fluorobenzoate, methyl-2-fluorobenzoate, 2-fluorobenzoic acid methyl ester, benzoic acid, 2-fluoro-, methyl ester, o-fluorobenzoic acid, methyl ester, benzoic acid, o-fluoro-, methyl ester, 2-fluoro-benzoic acid methyl ester, methyl2-fluorobenzoate, pubchem3565, acmc-209j5e
PubChem CID 67854
IUPAC Name methyl 2-fluorobenzoate
SMILES COC(=O)C1=CC=CC=C1F

Specifications

Color Colorless
Boiling Point 102°C
Quantity 25 g
Formula Weight 154.14
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl 2-Fluorobenzoate