Learn More
Methyl 2-fluoroacrylate, 95%, stab. with 1% BHT
CAS: 2343-89-7 | C4H5FO2 | 104.08 g/mol
$146.00 - $582.55
Chemical Identifiers
| CAS | 2343-89-7 |
|---|---|
| Molecular Formula | C4H5FO2 |
| Molecular Weight (g/mol) | 104.08 |
| MDL Number | MFCD04039286 |
| InChI Key | ZTZJVAOTIOAZGZ-UHFFFAOYSA-N |
| Synonym | methyl 2-fluoroacrylate, methyl-2-fluoroacrylate, 2-fluoroacrylic acid methyl ester, methyl fluoroacrylate, 2-propenoic acid, 2-fluoro-, methyl ester, methyl a-fluoroacrylate, pubchem12658, methyl alpha-fluoroacrylate, acmc-209g3b, ksc493a9d |
| PubChem CID | 2782524 |
| IUPAC Name | methyl 2-fluoroprop-2-enoate |
| SMILES | COC(=O)C(=C)F |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH6134306
|
Thermo Scientific Chemicals
H6134306 |
5 g |
Each for $146.00
|
|
|||||
|
AAH6134314
|
Thermo Scientific Chemicals
H6134314 |
25 g |
Each for $582.55
|
|
|||||
Description
Methyl 2-fluoroacrylate is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsMethyl 2-fluoroacrylate is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.
Solubility
Slightly soluble in water.
Notes
Store in cool, well ventilated area. Keep container tightly closed.
Chemical Identifiers
| 2343-89-7 | |
| 104.08 | |
| ZTZJVAOTIOAZGZ-UHFFFAOYSA-N | |
| 2782524 | |
| COC(=O)C(=C)F |
| C4H5FO2 | |
| MFCD04039286 | |
| methyl 2-fluoroacrylate, methyl-2-fluoroacrylate, 2-fluoroacrylic acid methyl ester, methyl fluoroacrylate, 2-propenoic acid, 2-fluoro-, methyl ester, methyl a-fluoroacrylate, pubchem12658, methyl alpha-fluoroacrylate, acmc-209g3b, ksc493a9d | |
| methyl 2-fluoroprop-2-enoate |
Specifications
| 2343-89-7 | |
| CH2=C(F)CO2CH3 | |
| 5 g | |
| methyl 2-fluoroacrylate, methyl-2-fluoroacrylate, 2-fluoroacrylic acid methyl ester, methyl fluoroacrylate, 2-propenoic acid, 2-fluoro-, methyl ester, methyl a-fluoroacrylate, pubchem12658, methyl alpha-fluoroacrylate, acmc-209g3b, ksc493a9d | |
| ZTZJVAOTIOAZGZ-UHFFFAOYSA-N | |
| methyl 2-fluoroprop-2-enoate | |
| 2782524 | |
| 95% | |
| Methyl 2-fluoroacrylate |
| C4H5FO2 | |
| MFCD04039286 | |
| UN1993 | |
| Slightly soluble in water. | |
| COC(=O)C(=C)F | |
| 104.08 | |
| 104.08 | |
| Liquid |
Safety and Handling
GHS H Statement
H224-H301-H315-H319-H335
Extremely flammable liquid and vapor.
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P201-P202-P210-P233-P240-P241-P242-P243-P260-P264b-P270-P281-P301+P310-P303+P361+P353-P308+P313-P330-P370+P378q-P501c
H225-H301-H360-H372
DOTInformation : Transport Hazard Class: 3; Packaging Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only