CAS RN 2343-89-7
IUPAC Name:
methyl 2-fluoroprop-2-enoate
Synonyms:
methyl 2-fluoroacrylate
Molecular Weight (g/mol):
104.08
Molecular Formula:
C4H5FO2
InChi Key:
ZTZJVAOTIOAZGZ-UHFFFAOYSA-N
SMILES:
COC(=O)C(F)=C
1
–
1
of
1
results
1
Promotions Available
Methyl 2-fluoroacrylate, 95%, stab. with 1% BHT
Methyl 2-fluoroacrylate, 95%, stab. with 1% BHT
CAS: 2343-89-7 Molecular Formula: C4H5FO2 Molecular Weight (g/mol): 104.08 MDL Number: MFCD04039286 InChI Key: ZTZJVAOTIOAZGZ-UHFFFAOYSA-N Synonym: methyl 2-fluoroacrylate,methyl-2-fluoroacrylate,2-fluoroacrylic acid methyl ester,methyl fluoroacrylate,2-propenoic acid, 2-fluoro-, methyl ester,methyl a-fluoroacrylate,pubchem12658,methyl alpha-fluoroacrylate,acmc-209g3b,ksc493a9d PubChem CID: 2782524 IUPAC Name: methyl 2-fluoroprop-2-enoate SMILES: COC(=O)C(=C)F