missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-fluoroacrylate, 95%, stab. with 1% BHT

Catalog No. AAH6134314
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6134314 25 g
AAH6134306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6134314 Supplier Thermo Scientific Chemicals Supplier No. H6134314
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2343-89-7 | C4H5FO2 | 104.08 g/mol

Methyl 2-fluoroacrylate is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 2-fluoroacrylate is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

Solubility
Slightly soluble in water.

Notes
Store in cool, well ventilated area. Keep container tightly closed.

Chemical Identifiers

CAS 2343-89-7
Molecular Formula C4H5FO2
Molecular Weight (g/mol) 104.08
MDL Number MFCD04039286
InChI Key ZTZJVAOTIOAZGZ-UHFFFAOYSA-N
Synonym methyl 2-fluoroacrylate, methyl-2-fluoroacrylate, 2-fluoroacrylic acid methyl ester, methyl fluoroacrylate, 2-propenoic acid, 2-fluoro-, methyl ester, methyl a-fluoroacrylate, pubchem12658, methyl alpha-fluoroacrylate, acmc-209g3b, ksc493a9d
PubChem CID 2782524
IUPAC Name methyl 2-fluoroprop-2-enoate
SMILES COC(=O)C(=C)F

Specifications

Linear Formula CH2=C(F)CO2CH3
Quantity 25 g
UN Number UN1993
Solubility Information Slightly soluble in water.
Formula Weight 104.08
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Methyl 2-fluoroacrylate
Hazard Category H225-H301-H360-H372
Hazard Statement GHS H Statement
H224-H301-H315-H319-H335
Extremely flammable liquid and vapor.
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P201-P202-P210-P233-P240-P241-P242-P243-P260-P264b-P270-P281-P301+P310-P303+P361+P353-P308+P313-P330-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packaging Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only