missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-(Cyanomethyl)benzoate 98.0+%, TCI America™

Catalog No. C15525G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C15525G 5 g
C155225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C15525G Supplier TCI America Supplier No. C15525G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 4-6-5597
Molecular Formula C10H9NO2
Molecular Weight (g/mol) 175.19
MDL Number MFCD01631304
InChI Key CMADSXVGXNLKHA-UHFFFAOYSA-N
Synonym 2-(Cyanomethyl)benzoic Acid Methyl Ester
PubChem CID 588805
IUPAC Name methyl 2-(cyanomethyl)benzoate
SMILES COC(=O)C1=CC=CC=C1CC#N

Specifications

Melting Point 49°C
Color White-Yellow
Boiling Point 133°C
Quantity 5 g
UN Number 3439
Formula Weight 175.19
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 2-(Cyanomethyl)benzoate