missing translation for 'onlineSavingsMsg'
Learn More

Methyl (2-Chlorophenyl)acetate 98.0+%, TCI America™

Catalog No. p-7125148
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C174025G 25 g
C1740100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C174025G Supplier TCI America Supplier No. C174025G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 57486-68-7
Molecular Formula C9H9ClO2
Molecular Weight (g/mol) 184.619
MDL Number MFCD00137445
InChI Key KHBWTRFWQROKJZ-UHFFFAOYSA-N
Synonym methyl 2-chlorophenylacetate, methyl 2-2-chlorophenyl acetate, methyl 2-chlorophenyl acetate, 2-chlorophenyl acetic acid methyl ester, benzeneacetic acid, 2-chloro-, methyl ester, 2-chlorophenylacetic acid methylester, 2-chlorobenzeneacetic acid methyl ester, acmc-209lyj, methyl2-chlorophenylacetate
PubChem CID 93683
IUPAC Name methyl 2-(2-chlorophenyl)acetate
SMILES COC(=O)CC1=CC=CC=C1Cl

Specifications

Color Yellow
Quantity 25 g
Physical Form Clear Liquid at 20°C
Boiling Point 128°C
Formula Weight 184.62
Percent Purity ≥98.0% (GC)
Chemical Name or Material Methyl (2-Chlorophenyl)acetate