accessibility menu, dialog, popup

UserName

CAS RN 57486-68-7

IUPAC Name: methyl 2-(2-chlorophenyl)acetate
Synonyms: methyl 2-(2-chlorophenyl)acetate
Molecular Weight (g/mol): 184.62
Molecular Formula: C9H9ClO2
InChi Key: KHBWTRFWQROKJZ-UHFFFAOYSA-N
SMILES: COC(=O)CC1=CC=CC=C1Cl
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)