missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 2-(Chloromethyl)benzoate 97.0+%, TCI America™
$298.25 - $1191.13
Chemical Identifiers
| CAS | 34040-62-5 |
|---|---|
| Molecular Formula | C9H9ClO2 |
| Molecular Weight (g/mol) | 184.62 |
| MDL Number | MFCD06200781 |
| InChI Key | AAHZCIWUDPKSJP-UHFFFAOYSA-N |
| Synonym | 2-(Chloromethyl)benzoic Acid Methyl Ester |
| PubChem CID | 118590 |
| IUPAC Name | methyl 2-(chloromethyl)benzoate |
| SMILES | COC(=O)C1=CC=CC=C1CCl |
Chemical Identifiers
| 34040-62-5 | |
| 184.62 | |
| AAHZCIWUDPKSJP-UHFFFAOYSA-N | |
| 118590 | |
| COC(=O)C1=CC=CC=C1CCl |
| C9H9ClO2 | |
| MFCD06200781 | |
| 2-(Chloromethyl)benzoic Acid Methyl Ester | |
| methyl 2-(chloromethyl)benzoate |
Specifications
| 34040-62-5 | |
| C9H9ClO2 | |
| 5 g | |
| 2-(Chloromethyl)benzoic Acid Methyl Ester | |
| COC(=O)C1=CC=CC=C1CCl | |
| 184.62 | |
| 184.62 | |
| Liquid |
| Yellow | |
| MFCD06200781 | |
| 3265 | |
| AAHZCIWUDPKSJP-UHFFFAOYSA-N | |
| methyl 2-(chloromethyl)benzoate | |
| 118590 | |
| ≥97.0% (GC) | |
| Methyl 2-(Chloromethyl)benzoate |
Safety and Handling
TSCA : No