missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 2-(Chloromethyl)benzoate 97.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Quantity | 5 g |
| UN Number | 3265 |
| Formula Weight | 184.62 |
| Percent Purity | ≥97.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | Methyl 2-(Chloromethyl)benzoate |
Safety and Handling
| TSCA | No |