missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 2-Chloro-D-mandelate 98.0+%, TCI America™
Supplier: TCI America M238225G
Specifications
| Methyl 2-Chloro-D-mandelate | |
| 32345-59-8 | |
| 25 g | |
| C9H9ClO3 | |
| 2-Chloro-D-mandelic Acid Methyl Ester, Methyl (R)-2′-Chloro-alpha-hydroxyphenylacetate, (R)-2′-Chloro-alpha-hydroxyphenylacetic Acid Methyl Ester | |
| COC(=O)C(C1=CC=CC=C1Cl)O | |
| 200.618 | |
| 200.62 |
| 34°C | |
| White-Yellow | |
| Crystal-Lump at 20°C | |
| MFCD02259751 | |
| ZMPGBVQQIQSQED-MRVPVSSYSA-N | |
| methyl (2R)-2-(2-chlorophenyl)-2-hydroxyacetate | |
| 11790232 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 32345-59-8 | |
| 200.618 | |
| ZMPGBVQQIQSQED-MRVPVSSYSA-N | |
| methyl (2R)-2-(2-chlorophenyl)-2-hydroxyacetate |
| C9H9ClO3 | |
| MFCD02259751 | |
| 11790232 | |
| COC(=O)C(C1=CC=CC=C1Cl)O |
Safety and Handling
TSCA : No