missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-Chloro-D-mandelate 98.0+%, TCI America™

Catalog No. p-7133196
Change view
Click to view available options
Quantity:
25 g
5 g
Catalog No. Quantity
M23825G 5 g
M238225G 25 g
2 options

Catalog No. M23825G

Supplier: TCI America M23825G

Only null left

Chemical Identifiers

CAS 32345-59-8
Molecular Formula C9H9ClO3
Molecular Weight (g/mol) 200.618
MDL Number MFCD02259751
InChI Key ZMPGBVQQIQSQED-MRVPVSSYSA-N
PubChem CID 11790232
IUPAC Name methyl (2R)-2-(2-chlorophenyl)-2-hydroxyacetate
SMILES COC(=O)C(C1=CC=CC=C1Cl)O

Specifications

Melting Point 34°C
Color White-Yellow
Quantity 5 g
Physical Form Crystal-Lump at 20°C
Synonym 2-Chloro-D-mandelic Acid Methyl Ester, Methyl (R)-2′C-Chloro-alpha-hydroxyphenylacetate, (R)-2′C-Chloro-alpha-hydroxyphenylacetic Acid Methyl Ester
Formula Weight 200.62
Percent Purity ≥98.0% (GC)
Chemical Name or Material Methyl 2-Chloro-D-mandelate