missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 2-Chloro-4-nitrobenzoate 99.0+%, TCI America™
$105.89 - $302.25
Chemical Identifiers
| CAS | 13324-11-3 |
|---|---|
| Molecular Formula | C8H6ClNO4 |
| Molecular Weight (g/mol) | 215.59 |
| MDL Number | MFCD00017013 |
| InChI Key | PICNSXCJRMYANX-UHFFFAOYSA-N |
| Synonym | 2-chloro-4-nitrobenzoic acid methyl ester, methyl2-chloro-4-nitrobenzoate, 4-nitro-2-chlorobenzoic acid methyl ester, 2-chloro-4-nitro-benzoic acid methyl ester, benzoic acid, 2-chloro-4-nitro-, methyl ester, maybridge1_005088, acmc-1c12v, methyl-2-chloro-4-nitrobenzoate, methyl-4-nitro-2-chlorobenzoate, 2-chloro-4-nitrobenzoic acid methylester |
| PubChem CID | 83343 |
| IUPAC Name | methyl 2-chloro-4-nitrobenzoate |
| SMILES | COC(=O)C1=CC=C(C=C1Cl)[N+]([O-])=O |
Chemical Identifiers
| 13324-11-3 | |
| 215.59 | |
| PICNSXCJRMYANX-UHFFFAOYSA-N | |
| 83343 | |
| COC(=O)C1=CC=C(C=C1Cl)[N+]([O-])=O |
| C8H6ClNO4 | |
| MFCD00017013 | |
| 2-chloro-4-nitrobenzoic acid methyl ester, methyl2-chloro-4-nitrobenzoate, 4-nitro-2-chlorobenzoic acid methyl ester, 2-chloro-4-nitro-benzoic acid methyl ester, benzoic acid, 2-chloro-4-nitro-, methyl ester, maybridge1_005088, acmc-1c12v, methyl-2-chloro-4-nitrobenzoate, methyl-4-nitro-2-chlorobenzoate, 2-chloro-4-nitrobenzoic acid methylester | |
| methyl 2-chloro-4-nitrobenzoate |
Specifications
| 13324-11-3 | |
| White-Yellow | |
| MFCD00017013 | |
| 2-chloro-4-nitrobenzoic acid methyl ester, methyl2-chloro-4-nitrobenzoate, 4-nitro-2-chlorobenzoic acid methyl ester, 2-chloro-4-nitro-benzoic acid methyl ester, benzoic acid, 2-chloro-4-nitro-, methyl ester, maybridge1_005088, acmc-1c12v, methyl-2-chloro-4-nitrobenzoate, methyl-4-nitro-2-chlorobenzoate, 2-chloro-4-nitrobenzoic acid methylester | |
| COC(=O)C1=CC=C(C=C1Cl)[N+]([O-])=O | |
| 215.59 | |
| 215.59 | |
| Crystalline Powder |
| 77°C | |
| C8H6ClNO4 | |
| 5 g | |
| PICNSXCJRMYANX-UHFFFAOYSA-N | |
| methyl 2-chloro-4-nitrobenzoate | |
| 83343 | |
| ≥99.0% (GC) | |
| Methyl 2-Chloro-4-nitrobenzoate |
Safety and Handling
TSCA : No