missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-Chloro-4-nitrobenzoate 99.0+%, TCI America™

Catalog No. M17245G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M17245G 5 g
M172425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M17245G Supplier TCI America Supplier No. M17245G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 13324-11-3
Molecular Formula C8H6ClNO4
Molecular Weight (g/mol) 215.59
MDL Number MFCD00017013
InChI Key PICNSXCJRMYANX-UHFFFAOYSA-N
Synonym 2-chloro-4-nitrobenzoic acid methyl ester, methyl2-chloro-4-nitrobenzoate, 4-nitro-2-chlorobenzoic acid methyl ester, 2-chloro-4-nitro-benzoic acid methyl ester, benzoic acid, 2-chloro-4-nitro-, methyl ester, maybridge1_005088, acmc-1c12v, methyl-2-chloro-4-nitrobenzoate, methyl-4-nitro-2-chlorobenzoate, 2-chloro-4-nitrobenzoic acid methylester
PubChem CID 83343
IUPAC Name methyl 2-chloro-4-nitrobenzoate
SMILES COC(=O)C1=CC=C(C=C1Cl)[N+]([O-])=O

Specifications

Melting Point 77°C
Color White-Yellow
Quantity 5 g
Formula Weight 215.59
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 2-Chloro-4-nitrobenzoate