missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-chloro-4-nitrobenzoate, 98%

Catalog No. AAB2016922
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2016922 100 g
AAB2016906 5 g
AAB2016914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2016922 Supplier Thermo Scientific Chemicals Supplier No. B2016922
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 13324-11-3 | C8H6ClNO4 | 215.59 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13324-11-3
Molecular Formula C8H6ClNO4
Molecular Weight (g/mol) 215.59
MDL Number MFCD00017013
InChI Key PICNSXCJRMYANX-UHFFFAOYSA-N
Synonym 2-chloro-4-nitrobenzoic acid methyl ester, methyl2-chloro-4-nitrobenzoate, 4-nitro-2-chlorobenzoic acid methyl ester, 2-chloro-4-nitro-benzoic acid methyl ester, benzoic acid, 2-chloro-4-nitro-, methyl ester, maybridge1_005088, acmc-1c12v, methyl-2-chloro-4-nitrobenzoate, methyl-4-nitro-2-chlorobenzoate, 2-chloro-4-nitrobenzoic acid methylester
PubChem CID 83343
IUPAC Name methyl 2-chloro-4-nitrobenzoate
SMILES COC(=O)C1=CC=C(C=C1Cl)[N+]([O-])=O

Specifications

Melting Point 74°C to 78°C
Quantity 100 g
Beilstein 1877723
Formula Weight 215.59
Percent Purity 98%
Chemical Name or Material Methyl 2-chloro-4-nitrobenzoate
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 236-362-3
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only