missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-Bromothiazole-4-carboxylate 98.0+%, TCI America™

Catalog No. m26151g
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
5 g
Catalog No. Quantity
M26151G 1 g
M26155G 5 g
2 options

Catalog No. M26151G

Supplier: TCI America M26151G

Only null left

Chemical Identifiers

CAS 170235-26-4
Molecular Formula C5H4BrNO2S
Molecular Weight (g/mol) 222.056
MDL Number MFCD06659908
InChI Key YOWKNNKTQWCYNC-UHFFFAOYSA-N
PubChem CID 2763213
IUPAC Name methyl 2-bromo-1,3-thiazole-4-carboxylate
SMILES COC(=O)C1=CSC(=N1)Br

Specifications

Melting Point 130°C
Color Yellow
Quantity 1 g
Synonym methyl 2-bromothiazole-4-carboxylate, 2-bromothiazole-4-carboxylic acid methyl ester, 2-bromo-4-methoxycarbonyl-1,3-thiazole, methyl2-bromothiazole-4-carboxylate, methyl-2-bromothiazole-4-carboxylate, 4-thiazolecarboxylic acid, 2-bromo-, methyl ester, 2-bromo-thiazole-4-carboxylic acid methyl ester, 2-bromo-1,3-thiazole-4-carboxylic acid methyl ester, pubchem8920, acmc-1c5qt
Formula Weight 222.06
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 2-Bromothiazole-4-carboxylate