missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-bromothiazole-4-carboxylate, 96%, Thermo Scientific Chemicals

$402.49 - $402.49

Chemical Identifiers

CAS 170235-26-4
Molecular Formula C5H4BrNO2S
Molecular Weight (g/mol) 222.056
MDL Number MFCD06659908
InChI Key YOWKNNKTQWCYNC-UHFFFAOYSA-N
PubChem CID 2763213
IUPAC Name methyl 2-bromo-1,3-thiazole-4-carboxylate
SMILES COC(=O)C1=CSC(=N1)Br
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6354106
View Documents
Thermo Scientific Chemicals
H6354106
5 g
Each for $402.49
Only null left
 
Description

Description

It is employed as a intermediate for pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a intermediate for pharmaceutical.

Solubility
Slightly soluble in water.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications

Chemical Identifiers

170235-26-4
222.056
YOWKNNKTQWCYNC-UHFFFAOYSA-N
methyl 2-bromo-1,3-thiazole-4-carboxylate
C5H4BrNO2S
MFCD06659908
2763213
COC(=O)C1=CSC(=N1)Br

Specifications

170235-26-4
MFCD06659908
methyl 2-bromothiazole-4-carboxylate, 2-bromothiazole-4-carboxylic acid methyl ester, 2-bromo-4-methoxycarbonyl-1,3-thiazole, methyl2-bromothiazole-4-carboxylate, methyl-2-bromothiazole-4-carboxylate, 4-thiazolecarboxylic acid, 2-bromo-, methyl ester, 2-bromo-thiazole-4-carboxylic acid methyl ester, 2-bromo-1,3-thiazole-4-carboxylic acid methyl ester, pubchem8920, acmc-1c5qt
YOWKNNKTQWCYNC-UHFFFAOYSA-N
methyl 2-bromo-1,3-thiazole-4-carboxylate
2763213
96%
Methyl 2-bromothiazole-4-carboxylate
C5H4BrNO2S
5 g
Slightly soluble in water.
COC(=O)C1=CSC(=N1)Br
222.056
222.06
Solid
Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only