missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 2-Bromoisonicotinate 98.0+%, TCI America™
$203.17 - $609.54
Chemical Identifiers
| CAS | 26156-48-9 |
|---|---|
| Molecular Formula | C7H6BrNO2 |
| Molecular Weight (g/mol) | 216.034 |
| MDL Number | MFCD03791265 |
| InChI Key | MULLTHQTADMZDM-UHFFFAOYSA-N |
| Synonym | 2-Bromoisonicotinic Acid Methyl Ester, Methyl 2-Bromopyridine-4-carboxylate, 2-Bromopyridine-4-carboxylic Acid Methyl Ester |
| PubChem CID | 3847768 |
| IUPAC Name | methyl 2-bromopyridine-4-carboxylate |
| SMILES | COC(=O)C1=CC(=NC=C1)Br |
Chemical Identifiers
| 26156-48-9 | |
| 216.034 | |
| MULLTHQTADMZDM-UHFFFAOYSA-N | |
| 3847768 | |
| COC(=O)C1=CC(=NC=C1)Br |
| C7H6BrNO2 | |
| MFCD03791265 | |
| 2-Bromoisonicotinic Acid Methyl Ester, Methyl 2-Bromopyridine-4-carboxylate, 2-Bromopyridine-4-carboxylic Acid Methyl Ester | |
| methyl 2-bromopyridine-4-carboxylate |
Specifications
| 26156-48-9 | |
| White-Yellow | |
| MFCD03791265 | |
| 2-Bromoisonicotinic Acid Methyl Ester, Methyl 2-Bromopyridine-4-carboxylate, 2-Bromopyridine-4-carboxylic Acid Methyl Ester | |
| COC(=O)C1=CC(=NC=C1)Br | |
| 216.034 | |
| 216.03 | |
| Crystalline Powder |
| 37°C | |
| C7H6BrNO2 | |
| 1 g | |
| MULLTHQTADMZDM-UHFFFAOYSA-N | |
| methyl 2-bromopyridine-4-carboxylate | |
| 3847768 | |
| ≥98.0% (GC) | |
| Methyl 2-Bromoisonicotinate |
Safety and Handling
TSCA : No