CAS RN 26156-48-9
IUPAC Name:
methyl 2-bromopyridine-4-carboxylate
Synonyms:
methyl 2-bromopyridine-4-carboxylate
Molecular Weight (g/mol):
216.03
Molecular Formula:
C7H6BrNO2
InChi Key:
MULLTHQTADMZDM-UHFFFAOYSA-N
SMILES:
COC(=O)C1=CC=NC(Br)=C1
1
–
1
of
1
results
1
Methyl 2-Bromoisonicotinate 98.0+%, TCI America™
CAS: 26156-48-9 Molecular Formula: C7H6BrNO2 Molecular Weight (g/mol): 216.034 MDL Number: MFCD03791265 InChI Key: MULLTHQTADMZDM-UHFFFAOYSA-N Synonym: 2-Bromoisonicotinic Acid Methyl Ester, Methyl 2-Bromopyridine-4-carboxylate, 2-Bromopyridine-4-carboxylic Acid Methyl Ester PubChem CID: 3847768 IUPAC Name: methyl 2-bromopyridine-4-carboxylate SMILES: COC(=O)C1=CC(=NC=C1)Br