missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-Bromoisonicotinate 98.0+%, TCI America™

Catalog No. M24615G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M24615G 5 g
M24611G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M24615G Supplier TCI America Supplier No. M24615G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 26156-48-9 | C7H6BrNO2 | 216.034 g/mol

Chemical Identifiers

CAS 26156-48-9
Molecular Formula C7H6BrNO2
Molecular Weight (g/mol) 216.034
MDL Number MFCD03791265
InChI Key MULLTHQTADMZDM-UHFFFAOYSA-N
Synonym 2-Bromoisonicotinic Acid Methyl Ester, Methyl 2-Bromopyridine-4-carboxylate, 2-Bromopyridine-4-carboxylic Acid Methyl Ester
PubChem CID 3847768
IUPAC Name methyl 2-bromopyridine-4-carboxylate
SMILES COC(=O)C1=CC(=NC=C1)Br

Specifications

Melting Point 37°C
Color White-Yellow
Quantity 5 g
Formula Weight 216.03
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 2-Bromoisonicotinate