missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-Bromobenzoate 98.0+%, TCI America™

Catalog No. B055525G
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B055525G 25 g
B0555250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B055525G Supplier TCI America Supplier No. B055525G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 610-94-6
Molecular Formula C8H7BrO2
Molecular Weight (g/mol) 215.05
MDL Number MFCD00016328
InChI Key SWGQITQOBPXVRC-UHFFFAOYSA-N
Synonym methyl o-bromobenzoate, 2-bromobenzoic acid methyl ester, benzoic acid, 2-bromo-, methyl ester, methyl-2-bromobenzoate, methyl bromobenzoate, o-bromobenzoic acid, methyl ester, benzoic acid, o-bromo-, methyl ester, pubchem3956, 2-bromo methyl benzoate, acmc-1atow
PubChem CID 11894
IUPAC Name methyl 2-bromobenzoate
SMILES COC(=O)C1=CC=CC=C1Br

Specifications

Color Yellow
Boiling Point 253°C
Quantity 25 g
Formula Weight 215.05
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl 2-Bromobenzoate