missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-bromobenzoate, 99%

Catalog No. AAA1243514
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1243514 25 g
AAA1243522 100 g
AAA1243536 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1243514 Supplier Thermo Scientific Chemicals Supplier No. A1243514
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 610-94-6 | C8H7BrO2 | 215.05 g/mol

Methyl 2-bromobenzoate under Heck reaction with acrylonitrile results in lactonization to give the benzofuran-2-one 3-(cyanomethyl)-2-coumaranone. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 2-bromobenzoate under Heck reaction with acrylonitrile results in lactonization to give the benzofuran-2-one 3-(cyanomethyl)-2-coumaranone. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals

Solubility
Insoluble in water.

Notes
Store in a cool place. Keep away from oxidizing agents.

Chemical Identifiers

CAS 610-94-6
Molecular Formula C8H7BrO2
Molecular Weight (g/mol) 215.05
MDL Number MFCD00016328
InChI Key SWGQITQOBPXVRC-UHFFFAOYSA-N
Synonym methyl o-bromobenzoate, 2-bromobenzoic acid methyl ester, benzoic acid, 2-bromo-, methyl ester, methyl-2-bromobenzoate, methyl bromobenzoate, o-bromobenzoic acid, methyl ester, benzoic acid, o-bromo-, methyl ester, pubchem3956, 2-bromo methyl benzoate, acmc-1atow
PubChem CID 11894
IUPAC Name methyl 2-bromobenzoate
SMILES COC(=O)C1=CC=CC=C1Br

Specifications

Density 1.532
Boiling Point 252°C
Flash Point >110°C (230°F)
Refractive Index 1.559
Quantity 25 g
Beilstein 1862501
Solubility Information Insoluble in water.
Formula Weight 215.05
Percent Purity 99%
Chemical Name or Material Methyl 2-bromobenzoate
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 210-241-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only