missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 2-Bromo-5-nitrobenzoate 98.0+%, TCI America™
$68.97 - $197.59
Chemical Identifiers
| CAS | 6942-36-5 |
|---|---|
| Molecular Formula | C8H6BrNO4 |
| Molecular Weight (g/mol) | 260.043 |
| MDL Number | MFCD00010867 |
| InChI Key | VSEYYEKRZNRECT-UHFFFAOYSA-N |
| Synonym | 2-bromo-5-nitrobenzoic acid methyl ester, benzoic acid, 2-bromo-5-nitro-, methyl ester, methyl 2-bromo-5-nitro-benzoate, 2-bromo-5-nitro-benzoic acid methyl ester, methyl2-bromo-5-nitrobenzoate, pubchem3971, acmc-1b9le, ksc495k5r, methyl 5-nitro-2-bromobenzoate, methyl-2-bromo-5-nitrobenzoate |
| PubChem CID | 245494 |
| IUPAC Name | methyl 2-bromo-5-nitrobenzoate |
| SMILES | COC(=O)C1=C(C=CC(=C1)[N+](=O)[O-])Br |
Chemical Identifiers
| 6942-36-5 | |
| 260.043 | |
| VSEYYEKRZNRECT-UHFFFAOYSA-N | |
| 245494 | |
| COC(=O)C1=C(C=CC(=C1)[N+](=O)[O-])Br |
| C8H6BrNO4 | |
| MFCD00010867 | |
| 2-bromo-5-nitrobenzoic acid methyl ester, benzoic acid, 2-bromo-5-nitro-, methyl ester, methyl 2-bromo-5-nitro-benzoate, 2-bromo-5-nitro-benzoic acid methyl ester, methyl2-bromo-5-nitrobenzoate, pubchem3971, acmc-1b9le, ksc495k5r, methyl 5-nitro-2-bromobenzoate, methyl-2-bromo-5-nitrobenzoate | |
| methyl 2-bromo-5-nitrobenzoate |
Specifications
| 6942-36-5 | |
| Yellow | |
| MFCD00010867 | |
| 2-bromo-5-nitrobenzoic acid methyl ester, benzoic acid, 2-bromo-5-nitro-, methyl ester, methyl 2-bromo-5-nitro-benzoate, 2-bromo-5-nitro-benzoic acid methyl ester, methyl2-bromo-5-nitrobenzoate, pubchem3971, acmc-1b9le, ksc495k5r, methyl 5-nitro-2-bromobenzoate, methyl-2-bromo-5-nitrobenzoate | |
| COC(=O)C1=C(C=CC(=C1)[N+](=O)[O-])Br | |
| 260.043 | |
| 260.04 | |
| Crystalline Powder |
| 82°C | |
| C8H6BrNO4 | |
| 5 g | |
| VSEYYEKRZNRECT-UHFFFAOYSA-N | |
| methyl 2-bromo-5-nitrobenzoate | |
| 245494 | |
| ≥98.0% (GC) | |
| Methyl 2-Bromo-5-nitrobenzoate |
Safety and Handling
TSCA : No