missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-bromo-5-nitrobenzoate, 98+%

Catalog No. AAB2128806
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2128806 5 g
AAB2128814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2128806 Supplier Thermo Scientific Chemicals Supplier No. B2128806
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 6942-36-5 | C8H6BrNO4 | 260.043 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6942-36-5
Molecular Formula C8H6BrNO4
Molecular Weight (g/mol) 260.043
MDL Number MFCD00010867
InChI Key VSEYYEKRZNRECT-UHFFFAOYSA-N
Synonym 2-bromo-5-nitrobenzoic acid methyl ester, benzoic acid, 2-bromo-5-nitro-, methyl ester, methyl 2-bromo-5-nitro-benzoate, 2-bromo-5-nitro-benzoic acid methyl ester, methyl2-bromo-5-nitrobenzoate, pubchem3971, acmc-1b9le, ksc495k5r, methyl 5-nitro-2-bromobenzoate, methyl-2-bromo-5-nitrobenzoate
PubChem CID 245494
IUPAC Name methyl 2-bromo-5-nitrobenzoate
SMILES COC(=O)C1=C(C=CC(=C1)[N+](=O)[O-])Br

Specifications

Melting Point 79°C to 82°C
Quantity 5 g
Beilstein 2617575
Formula Weight 260.05
Percent Purity ≥98%
Chemical Name or Material Methyl 2-bromo-5-nitrobenzoate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only