missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-Bromo-4-nitrobenzoate 98.0+%, TCI America™

Catalog No. m21231g
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M21231G 1 g
M21235G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M21231G Supplier TCI America Supplier No. M21231G

Chemical Identifiers

CAS 100959-22-6
Molecular Formula C8H6BrNO4
Molecular Weight (g/mol) 260.043
MDL Number MFCD07779271
InChI Key XYMZAFDNPJLOTP-UHFFFAOYSA-N
Synonym 2-Bromo-4-nitrobenzoic Acid Methyl Ester
PubChem CID 15833211
IUPAC Name methyl 2-bromo-4-nitrobenzoate
SMILES COC(=O)C1=C(C=C(C=C1)[N+](=O)[O-])Br

Specifications

Melting Point 84°C
Color Yellow
Quantity 1 g
Formula Weight 260.04
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 2-Bromo-4-nitrobenzoate