missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-Bromo-3-methylbenzoate 98.0+%, TCI America™

Catalog No. p-7133286
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M25465G 5 g
M254625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M25465G Supplier TCI America Supplier No. M25465G
Only null left

Chemical Identifiers

CAS 131001-86-0
Molecular Formula C9H9BrO2
Molecular Weight (g/mol) 229.07
MDL Number MFCD00144762
InChI Key QAOFGUXVDAZKBW-UHFFFAOYSA-N
Synonym 2-bromo-3-methyl-benzoic acid methyl ester, methyl2-bromo-3-methylbenzoate, benzoic acid, 2-bromo-3-methyl-, methyl ester, 2-bromo-3-methylbenzoic acid methyl ester, methyl 2-bromo-3-methyl-benzoate, pubchem3966, acmc-1c3gk, methyl 2-bromo-m-toluate, intermediates-zcf02010, ksc495c9h
PubChem CID 10823279
IUPAC Name methyl 2-bromo-3-methylbenzoate
SMILES COC(=O)C1=C(Br)C(C)=CC=C1

Specifications

Color Yellow
Quantity 5 g
Formula Weight 229.07
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl 2-Bromo-3-methylbenzoate