missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-bromo-3-fluoropropionate, 97%

Catalog No. p-7030900
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH31790MD 250 mg
AAH3179003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH31790MD Supplier Thermo Scientific Chemicals Supplier No. H31790MD
Only null left

CAS: 1537-52-6 | C4H6BrFO2 | 184.99 g/mol

In organic synthesis. Employed as important intermediates in various areas such as peptide synthesis, asymmetric synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
In organic synthesis. Employed as important intermediates in various areas such as peptide synthesis, asymmetric synthesis.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, bases.

Chemical Identifiers

CAS 1537-52-6
Molecular Formula C4H6BrFO2
Molecular Weight (g/mol) 184.99
MDL Number MFCD09800641
InChI Key BXTYDNLOFHBZFS-UHFFFAOYNA-N
Synonym 2-bromo-3-fluoropropionic acid methyl ester, methyl 2-bromo-3-fluoropropionate, methyl 2-bromo-3-fluoro-propanoate, methyl alpha-bromo-beta-fluoropropionate, methyl 2-bromo-3-fluoroproprionate, propanoic acid, 2-bromo-3-fluoro-, methyl ester
PubChem CID 5271584
IUPAC Name methyl 2-bromo-3-fluoropropanoate
SMILES COC(=O)C(Br)CF

Specifications

Boiling Point 50°C to 54°C (8 mmHg)
Quantity 250 mg
UN Number UN3265
Solubility Information Insoluble in water.
Formula Weight 184.99
Percent Purity 97%
Chemical Name or Material Methyl 2-bromo-3-fluoropropionate
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: III
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only