accessibility menu, dialog, popup

UserName

CAS RN 1537-52-6

IUPAC Name: methyl 2-bromo-3-fluoropropanoate
Synonyms: methyl 2-bromo-3-fluoropropanoate
Molecular Weight (g/mol): 184.99
Molecular Formula: C4H6BrFO2
InChi Key: BXTYDNLOFHBZFS-UHFFFAOYNA-N
SMILES: COC(=O)C(Br)CF
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results