missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 2-Benzoylbenzoate 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 52°C |
| Color | White |
| Boiling Point | 352°C |
| Quantity | 25 g |
| Formula Weight | 240.26 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Methyl 2-Benzoylbenzoate |
Safety and Handling
| EINECSNumber | (4)-0267 |
| TSCA | Yes |