missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-(Aminosulfonyl)benzoate 98.0+%, TCI America™

Catalog No. p-7132885
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M186625G 25 g
M1866500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M186625G Supplier TCI America Supplier No. M186625G
Only null left

Chemical Identifiers

CAS 57683-71-3
Molecular Formula C8H9NO4S
Molecular Weight (g/mol) 215.22
MDL Number MFCD00009808
InChI Key VSOOBQALJVLTBH-UHFFFAOYSA-N
Synonym methyl 2-aminosulfonyl benzoate, 2-carbomethoxybenzenesulfonamide, 2-sulfamoylbenzoic acid methyl ester, 2-methoxycarbonyl benzene sulfonamide, methyl o-sulphamoylbenzoate, benzoic acid, 2-aminosulfonyl-, methyl ester, benzoic acid, 2-sulfamoyl-, methyl ester, 2-methoxycarbonylphenylsulfonamide, 2-methoxycarbonylbenzenesulfonamide, 2-methoxycarbonyl benzenesulphonamide
PubChem CID 42546
ChEBI CHEBI:83512
IUPAC Name methyl 2-sulfamoylbenzoate
SMILES COC(=O)C1=CC=CC=C1S(N)(=O)=O

Specifications

Melting Point 124°C
Color White-Yellow
Quantity 25 g
Formula Weight 215.22
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 2-(Aminosulfonyl)benzoate
EINECSNumber (3)-2110
RTECSNumber DH6830000
TSCA Yes