missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-amino-5-methoxybenzoate, 96%

Catalog No. p-7036415
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH54704MD 250 mg
AAH5470403 1 g
AAH5470406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH54704MD Supplier Thermo Scientific Chemicals Supplier No. H54704MD
Only null left

CAS: 2475-80-1 | C9H11NO3 | 181.19 g/mol

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

Solubility
Slightly Soluble (1.1 g/L) (25°C) in water.

Notes
Store at room temperature. Keep away from oxidizing agents and acids. Moisture sensitive.

Chemical Identifiers

CAS 2475-80-1
Molecular Formula C9H11NO3
Molecular Weight (g/mol) 181.19
MDL Number MFCD09038482
InChI Key MOVBJUGHBJJKOW-UHFFFAOYSA-N
PubChem CID 13052275
IUPAC Name methyl 2-amino-5-methoxybenzoate
SMILES COC(=O)C1=C(N)C=CC(OC)=C1

Specifications

Melting Point 32°C to 33°C
Quantity 250 mg
Synonym benzoic acid, 2-amino-5-methoxy-, methyl ester, methyl2-amino-5-methoxybenzoate, 2-amino-5-methoxybenzoic acid methyl ester, methyl 5-methoxyanthranilate, benzoic acid,2-amino-5-methoxy-, methyl ester, methyl 6-amino-m-anisate, acmc-1cnv1, intermediates-zcf02665, 2-amino-5-methoxybenzoic acid,methyl ester, 2-amino-5-methoxy-benzoic acid methyl ester
Solubility Information Slightly Soluble (1.1g/L) (25°C) in water.
Formula Weight 181.19
Percent Purity 96%
Chemical Name or Material Methyl 2-amino-5-methoxybenzoate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only