missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-amino-5-fluorobenzoate, 98%

Catalog No. AAL1977814
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1977814 25 g
AAL1977806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1977814 Supplier Thermo Scientific Chemicals Supplier No. L1977814
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 319-24-4 | C8H8FNO2 | 169.155 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 319-24-4
Molecular Formula C8H8FNO2
Molecular Weight (g/mol) 169.155
MDL Number MFCD03791308
InChI Key PUDDYSBKCDKATP-UHFFFAOYSA-N
Synonym 2-amino-5-fluorobenzoic acid methyl ester, methyl2-amino-5-fluorobenzoate, methyl 5-fluoroanthranilate, methyl 2-amino-5-fluoro-benzoate, benzoic acid, 2-amino-5-fluoro-, methyl ester, 2-amino-5-fluoro benzoic acid methyl ester, 2-amino-5-fluoro-benzoic acid methyl ester, pubchem3558, acmc-209hpn, ksc495g7j
PubChem CID 2783401
IUPAC Name methyl 2-amino-5-fluorobenzoate
SMILES COC(=O)C1=C(C=CC(=C1)F)N

Specifications

Melting Point 21°C to 25°C
Density 1.26
Boiling Point 100°C to 102°C (0.5 mmHg)
Flash Point 132°C (269°F)
Quantity 25 g
Beilstein 2803210
Formula Weight 169.16
Percent Purity 98%
Chemical Name or Material Methyl 2-amino-5-fluorobenzoate
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only