missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-amino-5-bromobenzoate, 98%

Catalog No. p-7023274
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1666606 5 g
AAA1666614 25 g
AAA1666622 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1666606 Supplier Thermo Scientific Chemicals Supplier No. A1666606

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 52727-57-8 | C8H8BrNO2 | 230.06 g/mol

Methyl 2-amino-5-bromobenzoate acts as a potential PqsD inhibitor against pseudomonas aeruginosa infections. It is also used in the preparation of hepatitis C virus NS5b RNA polymerase inhibitors. It is involved in plant growth-regulator activity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 2-amino-5-bromobenzoate acts as a potential PqsD inhibitor against pseudomonas aeruginosa infections. It is also used in the preparation of hepatitis C virus NS5b RNA polymerase inhibitors. It is involved in plant growth-regulator activity.

Solubility
Soluble in water.

Notes
Light and air sensitive. Incompatible with strong oxidizing agents, strong reducing agents, acid halides, acid anhydrides, strong acids and strong bases. Store in a cool place.

Chemical Identifiers

CAS 52727-57-8
Molecular Formula C8H8BrNO2
Molecular Weight (g/mol) 230.06
MDL Number MFCD00144761
InChI Key QVNYNHCNNGKULA-UHFFFAOYSA-N
Synonym methyl 5-bromoanthranilate, 2-amino-5-bromobenzoic acid methyl ester, 5-bromoanthranilic acid methyl ester, 2-amino-5-bromo-benzoic acid methyl ester, methyl 5-bromo-2-aminobenzoate, benzoic acid, 2-amino-5-bromo-, methyl ester, benzoic acid, 2-amino-5-bromo-, methyl ester, hydrochloride, pubchem3344, methyl-5-bromoanthranilate, acmc-1ats7
PubChem CID 736224
IUPAC Name methyl 2-amino-5-bromobenzoate
SMILES COC(=O)C1=CC(Br)=CC=C1N

Specifications

Melting Point 71°C to 73°C
Quantity 5 g
Beilstein 972720
Solubility Information Soluble in water.
Formula Weight 230.06
Percent Purity 98%
Chemical Name or Material Methyl 2-amino-5-bromobenzoate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only