missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-amino-4-chlorobenzoate, 98%

Catalog No. AAL1112509
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1112509 10 g
AAL1112518 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1112509 Supplier Thermo Scientific Chemicals Supplier No. L1112509
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 5900-58-3 | C8H8ClNO2 | 185.607 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5900-58-3
Molecular Formula C8H8ClNO2
Molecular Weight (g/mol) 185.607
MDL Number MFCD00017568
InChI Key YPSSCICDVDOEAI-UHFFFAOYSA-N
Synonym methyl 4-chloroanthranilate, 2-amino-4-chloro-benzoic acid methyl ester, methyl2-amino-4-chlorobenzoate, benzoic acid, 2-amino-4-chloro-, methyl ester, 2-amino-4-chlorobenzoic acid methyl ester, pubchem4643, methyl 4-chloro-anthranilate, methyl 2-amino4-chlorobenzoate, methyl-2-amino-4-chlorobenzoate
PubChem CID 80001
IUPAC Name methyl 2-amino-4-chlorobenzoate
SMILES COC(=O)C1=C(C=C(C=C1)Cl)N

Specifications

Melting Point 66°C to 68°C
Quantity 10 g
Beilstein 1072895
Formula Weight 185.61
Percent Purity 98%
Chemical Name or Material Methyl 2-amino-4-chlorobenzoate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 227-593-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only