missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-amino-4,5-dimethoxybenzoate, 98%

Catalog No. AAB2100218
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2100218 50 g
AAB2100209 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2100218 Supplier Thermo Scientific Chemicals Supplier No. B2100218
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 26759-46-6 | C10H13NO4 | 211.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 26759-46-6
Molecular Formula C10H13NO4
Molecular Weight (g/mol) 211.22
MDL Number MFCD00014904
InChI Key QQFHCCQSCQBKBG-UHFFFAOYSA-N
Synonym methyl 6-aminoveratrate, methyl 4,5-dimethoxyanthranilate, methyl 2-amino-4,5-dimethoxy benzoate, 2-amino-4,5-dimethoxybenzoic acid methyl ester, 6-aminoveratric acid methyl ester, methyl2-amino-4,5-dimethoxybenzoate, 4,5-dimethoxyanthranilic acid methyl, benzoic acid, 2-amino-4,5-dimethoxy-, methyl ester, methyl-2-amino-4,5-dimethoxybenzoate, 2-amino-4,5-dimethoxy-benzoic acid methyl ester
PubChem CID 611144
IUPAC Name methyl 2-amino-4,5-dimethoxybenzoate
SMILES COC(=O)C1=CC(OC)=C(OC)C=C1N

Specifications

Melting Point 128°C to 130°C
Quantity 50 g
Beilstein 2724754
Formula Weight 211.22
Percent Purity 98%
Chemical Name or Material Methyl 2-amino-4,5-dimethoxybenzoate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 247-967-7
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only