missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-Acetamido-5-chlorobenzoate 98.0+%, TCI America™

Catalog No. A160925G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A160925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A160925G Supplier TCI America Supplier No. A160925G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 20676-54-4
Molecular Formula C10H10ClNO3
Molecular Weight (g/mol) 227.64
MDL Number MFCD00144759
InChI Key TVAAIYFBEWHVCV-UHFFFAOYSA-N
Synonym 2-acetamido-5-chlorobenzoic acid methyl ester, methyl 5-chloro-2-acetamidobenzoate, methyl n-acetyl-5-chloroanthranilate, n-acetyl-5-chloroanthranilic acid methyl ester, 2-acetylamino-5-chloro-benzoic acid methyl ester, methyl 2-acetylamino-5-chlorobenzoate, pubchem3337, acmc-1cioq, methyl2-acetamido-5-chlorobenzoate, methyl 2-acetamido-5-chloranyl-benzoate
PubChem CID 4640571
IUPAC Name methyl 5-chloro-2-acetamidobenzoate
SMILES COC(=O)C1=CC(Cl)=CC=C1NC(C)=O

Specifications

Color White
Quantity 25 g
Formula Weight 227.64
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 2-Acetamido-5-chlorobenzoate