missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2,6-difluorobenzoate, 97%

Catalog No. AAL0806306
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0806306 5 g
AAL0806314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0806306 Supplier Thermo Scientific Chemicals Supplier No. L0806306
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 13671-00-6 | C8H6F2O2 | 172.131 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13671-00-6
Molecular Formula C8H6F2O2
Molecular Weight (g/mol) 172.131
MDL Number MFCD00051778
InChI Key QNPFLTKQLFSKBY-UHFFFAOYSA-N
PubChem CID 518798
IUPAC Name methyl 2,6-difluorobenzoate
SMILES COC(=O)C1=C(C=CC=C1F)F

Specifications

Density 1.281
Boiling Point 203°C to 204°C
Flash Point 95°C (203°F)
Refractive Index 1.476
Quantity 5 g
Beilstein 1948618
Synonym 2,6-difluorobenzoic acid, methyl ester, benzoic acid, 2,6-difluoro-, methyl ester, acmc-1bxyv, ksc494g7r, methyl2,6-difluorobenzoate, methyl 2,6-difluorobenzoate, 2,6-difluorobenzoic acid methyl ester, 2-methoxycarbonyl-1,3-difluorobenzene
Formula Weight 172.13
Percent Purity 97%
Chemical Name or Material Methyl 2,6-difluorobenzoate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only