missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2,5-Dihydroxycinnamate 96.0+%, TCI America™

Catalog No. p-7133266
Change view
Click to view available options
Quantity:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M252010MG 10 mg
M2520100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M252010MG Supplier TCI America Supplier No. M252010MG
Only null left

Chemical Identifiers

CAS 63177-57-1
Molecular Formula C10H10O4
Molecular Weight (g/mol) 194.19
MDL Number MFCD00132932
InChI Key BQCNSTFWSKOWMA-GORDUTHDSA-N
Synonym methyl 2,5-dihydroxycinnamate, erbstatin analog, methyl 3-2,5-dihydroxyphenyl acrylate, 2,5-dihydroxycinnamic acid methyl ester, 2,4-dihydroxymethylcinnamate, 2,5-dihydroxymethylcinnamate, chembl17329, methyl 3-2,5-dihydroxyphenyl-2-propenoate, methyl 2e-3-2,5-dihydroxyphenyl prop-2-enoate, 2-propenoic acid, 3-2,5-dihydroxyphenyl-, methyl ester
PubChem CID 5353609
ChEBI CHEBI:84089
IUPAC Name methyl (2E)-3-(2,5-dihydroxyphenyl)prop-2-enoate
SMILES COC(=O)\C=C\C1=CC(O)=CC=C1O

Specifications

Melting Point 182°C
Color Yellow
Quantity 10 mg
Formula Weight 194.19
Percent Purity ≥96.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 2,5-Dihydroxycinnamate