missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 2,5-Dihydrothiophene-3-carboxylate 95.0+%, TCI America™
Supplier: TCI America D28825G
Specifications
| Methyl 2,5-Dihydrothiophene-3-carboxylate | |
| 36°C | |
| C6H8O2S | |
| 5 g | |
| RDHSPSSFHQGPDS-UHFFFAOYSA-N | |
| methyl 2,5-dihydrothiophene-3-carboxylate | |
| 11457755 | |
| ≥95.0% (GC) |
| 67488-46-4 | |
| White-Yellow | |
| MFCD04038417 | |
| 2,5-Dihydrothiophene-3-carboxylic Acid Methyl Ester | |
| COC(=O)C1=CCSC1 | |
| 144.188 | |
| 144.19 | |
| Crystalline Lumps |
Chemical Identifiers
| 67488-46-4 | |
| 144.188 | |
| RDHSPSSFHQGPDS-UHFFFAOYSA-N | |
| 11457755 | |
| COC(=O)C1=CCSC1 |
| C6H8O2S | |
| MFCD04038417 | |
| 2,5-Dihydrothiophene-3-carboxylic Acid Methyl Ester | |
| methyl 2,5-dihydrothiophene-3-carboxylate |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator