missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2,5-Dihydrothiophene-3-carboxylate 95.0+%, TCI America™

Catalog No. p-7127554
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D28821G 1 g
D28825G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D28821G Supplier TCI America Supplier No. D28821G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 67488-46-4
Molecular Formula C6H8O2S
Molecular Weight (g/mol) 144.188
MDL Number MFCD04038417
InChI Key RDHSPSSFHQGPDS-UHFFFAOYSA-N
Synonym 2,5-Dihydrothiophene-3-carboxylic Acid Methyl Ester
PubChem CID 11457755
IUPAC Name methyl 2,5-dihydrothiophene-3-carboxylate
SMILES COC(=O)C1=CCSC1

Specifications

Melting Point 36°C
Color White-Yellow
Quantity 1 g
Formula Weight 144.19
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material Methyl 2,5-Dihydrothiophene-3-carboxylate
TSCA No
Recommended Storage Refrigerator