missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2,5-Dichloronicotinate 98.0+%, TCI America™

Catalog No. p-7133405
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M2752200MG 200 mg
M27521G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M2752200MG Supplier TCI America Supplier No. M2752200MG
Only null left

Chemical Identifiers

CAS 67754-03-4
Molecular Formula C7H5Cl2NO2
Molecular Weight (g/mol) 206.022
MDL Number MFCD03844857
InChI Key CNPFXYWTMBTSSI-UHFFFAOYSA-N
Synonym 2,5-Dichloronicotinic Acid Methyl Ester, Methyl 2,5-Dichloropyridine-3-carboxylate, 2,5-Dichloropyridine-3-carboxylic Acid Methyl Ester
PubChem CID 2764318
IUPAC Name methyl 2,5-dichloropyridine-3-carboxylate
SMILES COC(=O)C1=CC(=CN=C1Cl)Cl

Specifications

Melting Point 44°C
Color White
Quantity 200 mg
Formula Weight 206.02
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 2,5-Dichloronicotinate