CAS RN 67754-03-4
IUPAC Name:
methyl 2,5-dichloropyridine-3-carboxylate
Synonyms:
methyl 2,5-dichloropyridine-3-carboxylate
Molecular Weight (g/mol):
206.02
Molecular Formula:
C7H5Cl2NO2
InChi Key:
CNPFXYWTMBTSSI-UHFFFAOYSA-N
SMILES:
COC(=O)C1=CC(Cl)=CN=C1Cl
1
–
1
of
1
results
1
Methyl 2,5-Dichloronicotinate 98.0+%, TCI America™
CAS: 67754-03-4 Molecular Formula: C7H5Cl2NO2 Molecular Weight (g/mol): 206.022 MDL Number: MFCD03844857 InChI Key: CNPFXYWTMBTSSI-UHFFFAOYSA-N Synonym: 2,5-Dichloronicotinic Acid Methyl Ester, Methyl 2,5-Dichloropyridine-3-carboxylate, 2,5-Dichloropyridine-3-carboxylic Acid Methyl Ester PubChem CID: 2764318 IUPAC Name: methyl 2,5-dichloropyridine-3-carboxylate SMILES: COC(=O)C1=CC(=CN=C1Cl)Cl