missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2,4-dihydroxybenzoate, 97%

Catalog No. AAB2475714
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2475714 25 g
AAB2475722 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2475714 Supplier Thermo Scientific Chemicals Supplier No. B2475714
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 2150-47-2 | C8H8O4 | 168.148 g/mol

Methyl 2,4-dihydroxybenzoate is used as a cosmetic agents. And it is used in synthesis of raw material for fine chemicals, organic intermediate, pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 2,4-dihydroxybenzoate is used as a cosmetic agents. And it is used in synthesis of raw material for fine chemicals, organic intermediate, pharmaceutical intermediate.

Solubility
Soluble in methanol.

Notes
Keep container tightly closed. Store away from oxidizing agents.

Chemical Identifiers

CAS 2150-47-2
Molecular Formula C8H8O4
Molecular Weight (g/mol) 168.148
MDL Number MFCD00002276
InChI Key IIFCLXHRIYTHPV-UHFFFAOYSA-N
Synonym 2,4-dihydroxybenzoic acid methyl ester, benzoic acid, 2,4-dihydroxy-, methyl ester, methyl beta-resorcylate, methyl-2,4-dihydroxybenzoate, unii-xe68bu3oyj, xe68bu3oyj, .beta.-resorcylic acid, methyl ester, beta-resorcylic acid methyl ester, methyl benzoate, 2,4-dihydroxy, methyl b-resorcylate
PubChem CID 16523
IUPAC Name methyl 2,4-dihydroxybenzoate
SMILES COC(=O)C1=C(C=C(C=C1)O)O

Specifications

Melting Point 116°C to 119°C
Quantity 25 g
Solubility Information Soluble in methanol.
Formula Weight 168.15
Percent Purity 97%
Chemical Name or Material Methyl 2,4-dihydroxybenzoate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 218-428-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only