missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2,4-Difluorobenzoate 98.0+%, TCI America™

Catalog No. p-7132887
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M18795G 5 g
M187925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M18795G Supplier TCI America Supplier No. M18795G
Only null left

Chemical Identifiers

CAS 106614-28-2
Molecular Formula C8H6F2O2
Molecular Weight (g/mol) 172.131
MDL Number MFCD03093799
InChI Key YQUHULOBTDYMAG-UHFFFAOYSA-N
Synonym 2,4-difluoro-benzoic acid methyl ester, 2,4-difluorobenzoic acid methyl ester, methyl-2,4-difluorobenzoate, methyl 2,4-difluoro benzoate, benzoic acid, 2,4-difluoro-, methyl ester, pubchem15678, acmc-1c4sx, methyl 2,4-diflourobenzoate, ksc494g3p, rarechem al bf 0213
PubChem CID 2782897
IUPAC Name methyl 2,4-difluorobenzoate
SMILES COC(=O)C1=C(C=C(C=C1)F)F

Specifications

Color Yellow
Boiling Point 194°C
Quantity 5 g
Formula Weight 172.13
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl 2,4-Difluorobenzoate