Learn More
Methyl 2,4-dichloropyrimidine-6-carboxylate, 98%, Thermo Scientific™
Chemical Identifiers
| CAS | 6299-85-0 |
|---|---|
| Molecular Formula | C6H4Cl2N2O2 |
| Molecular Weight (g/mol) | 207.02 |
| InChI Key | DQNQNLWKAGZNIT-UHFFFAOYSA-N |
| Synonym | methyl 2,4-dichloropyrimidine-6-carboxylate, methyl 2,4-dichloro-pyrimidine-6-carboxylate, 2,6-dichloropyrimidine-4-carboxylic acid methyl ester, 4-pyrimidinecarboxylic acid, 2,6-dichloro-, methyl ester, 2,6-dichloro-4-pyrimidinecarboxylicacid methyl ester, t6n cnj bg dvo1 fg wln, zlchem 33, pubchem23452, methyl2,6-dichloropyrimidine-4-carboxylate, 2,4-dichloro-6-methoxycarbonyl pyrimidine |
| PubChem CID | 239689 |
| IUPAC Name | methyl 2,6-dichloropyrimidine-4-carboxylate |
| SMILES | COC(=O)C1=CC(=NC(=N1)Cl)Cl |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AC446580050
|
Thermo Scientific Chemicals
446580050 |
5g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC446580250
|
Thermo Scientific Chemicals
446580250 |
25g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 6299-85-0 | |
| 207.02 | |
| methyl 2,4-dichloropyrimidine-6-carboxylate, methyl 2,4-dichloro-pyrimidine-6-carboxylate, 2,6-dichloropyrimidine-4-carboxylic acid methyl ester, 4-pyrimidinecarboxylic acid, 2,6-dichloro-, methyl ester, 2,6-dichloro-4-pyrimidinecarboxylicacid methyl ester, t6n cnj bg dvo1 fg wln, zlchem 33, pubchem23452, methyl2,6-dichloropyrimidine-4-carboxylate, 2,4-dichloro-6-methoxycarbonyl pyrimidine | |
| methyl 2,6-dichloropyrimidine-4-carboxylate |
| C6H4Cl2N2O2 | |
| DQNQNLWKAGZNIT-UHFFFAOYSA-N | |
| 239689 | |
| COC(=O)C1=CC(=NC(=N1)Cl)Cl |
Specifications
| 6299-85-0 | |
| C6H4Cl2N2O2 | |
| methyl 2,4-dichloropyrimidine-6-carboxylate, methyl 2,4-dichloro-pyrimidine-6-carboxylate, 2,6-dichloropyrimidine-4-carboxylic acid methyl ester, 4-pyrimidinecarboxylic acid, 2,6-dichloro-, methyl ester, 2,6-dichloro-4-pyrimidinecarboxylicacid methyl ester, t6n cnj bg dvo1 fg wln, zlchem 33, pubchem23452, methyl2,6-dichloropyrimidine-4-carboxylate, 2,4-dichloro-6-methoxycarbonyl pyrimidine | |
| COC(=O)C1=CC(=NC(=N1)Cl)Cl | |
| 207.02 | |
| 207.02 | |
| Methyl 2, 4-dichloropyrimidine-6-carboxylate |
| 97.5% min. (GC) | |
| 5g | |
| DQNQNLWKAGZNIT-UHFFFAOYSA-N | |
| methyl 2,6-dichloropyrimidine-4-carboxylate | |
| 239689 | |
| 98% |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and ke
GHS Signal Word: Warning
RUO â Research Use Only