CAS RN 6299-85-0
IUPAC Name:
methyl 2,6-dichloropyrimidine-4-carboxylate
Synonyms:
methyl 2,6-dichloropyrimidine-4-carboxylate
Molecular Weight (g/mol):
207.01
Molecular Formula:
C6H4Cl2N2O2
InChi Key:
DQNQNLWKAGZNIT-UHFFFAOYSA-N
SMILES:
COC(=O)C1=CC(Cl)=NC(Cl)=N1
1
–
1
of
1
results
1
Promotions Available
Methyl 2,4-dichloropyrimidine-6-carboxylate, 98%
Methyl 2,4-dichloropyrimidine-6-carboxylate, 98%
CAS: 6299-85-0 Molecular Formula: C6H4Cl2N2O2 Molecular Weight (g/mol): 207.02 InChI Key: DQNQNLWKAGZNIT-UHFFFAOYSA-N Synonym: methyl 2,4-dichloropyrimidine-6-carboxylate,methyl 2,4-dichloro-pyrimidine-6-carboxylate,2,6-dichloropyrimidine-4-carboxylic acid methyl ester,4-pyrimidinecarboxylic acid, 2,6-dichloro-, methyl ester,2,6-dichloro-4-pyrimidinecarboxylicacid methyl ester,t6n cnj bg dvo1 fg wln,zlchem 33,pubchem23452,methyl2,6-dichloropyrimidine-4-carboxylate,2,4-dichloro-6-methoxycarbonyl pyrimidine PubChem CID: 239689 IUPAC Name: methyl 2,6-dichloropyrimidine-4-carboxylate SMILES: COC(=O)C1=CC(=NC(=N1)Cl)Cl