missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2,3-Dichloropropionate 95.0+%, TCI America™

Catalog No. D040810G
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D040810G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D040810G Supplier TCI America Supplier No. D040810G

Chemical Identifiers

CAS 3674-09-7
Molecular Formula C4H6Cl2O2
Molecular Weight (g/mol) 156.99
MDL Number MFCD00000944
InChI Key OFHMODDLBXETIK-UHFFFAOYNA-N
Synonym methyl 2,3-dichloropropionate, propanoic acid, 2,3-dichloro-, methyl ester, 2,3-dichloropropionic acid methyl ester, methyl-2,3-dichloropropionate, methyl alpha,beta-dichloropropionate, methyl .alpha.,.beta.-dichloropropionate, propionic acid, 2,3-dichloro-, methyl ester, propanoic acid,3-dichloro-, methyl ester, propionic acid,3-dichloro-, methyl ester, methyl-2,3-dichloropropioate
PubChem CID 95429
IUPAC Name methyl 2,3-dichloropropanoate
SMILES COC(=O)C(CCl)Cl

Specifications

Color Yellow
Boiling Point 82°C
Quantity 10 g
UN Number 1760
Formula Weight 156.99
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl 2,3-Dichloropropionate