accessibility menu, dialog, popup

UserName

H3COOClCl

CAS RN 3674-09-7

IUPAC Name: methyl 2,3-dichloropropanoate
Synonyms: methyl 2,3-dichloropropanoate
Molecular Weight (g/mol): 156.99
Molecular Formula: C4H6Cl2O2
InChi Key: OFHMODDLBXETIK-UHFFFAOYNA-N
SMILES: COC(=O)C(Cl)CCl
Boiling Point 
  • (1)
  • (3)
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)