missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2,3-Di-O-benzoyl-4,6-O-benzylidene-alpha-D-glucopyranoside 98.0+%, TCI America™

Catalog No. m20131g
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M20131G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M20131G Supplier TCI America Supplier No. M20131G

Chemical Identifiers

CAS 6748-91-0
Molecular Formula C28H26O8
Molecular Weight (g/mol) 490.508
MDL Number MFCD02167693
InChI Key CGMUHSNJRXPSSA-VBBSJTGDSA-N
PubChem CID 44629977
IUPAC Name [(4aS,6S,7S,8S,8aR)-7-benzoyloxy-6-methoxy-2-phenyl-4,4a,6,7,8,8a-hexahydropyrano[3,2-d][1,3]dioxin-8-yl] benzoate
SMILES COC1C(C(C2C(O1)COC(O2)C3=CC=CC=C3)OC(=O)C4=CC=CC=C4)OC(=O)C5=CC=CC=C5

Specifications

Melting Point 154°C
Color White
Quantity 1 g
Formula Weight 490.51
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 2,3-Di-O-benzoyl-4,6-O-benzylidene-alpha-D-glucopyranoside