accessibility menu, dialog, popup

UserName

CAS RN 6748-91-0

IUPAC Name: 7-(benzoyloxy)-6-methoxy-2-phenyl-hexahydro-2H-pyrano[3,2-d][1,3]dioxin-8-yl benzoate
Synonyms: 7-(benzoyloxy)-6-methoxy-2-phenyl-hexahydro-2H-pyrano[3,2-d][1,3]dioxin-8-yl benzoate
Molecular Weight (g/mol): 490.51
Molecular Formula: C28H26O8
InChi Key: CGMUHSNJRXPSSA-UHFFFAOYNA-N
SMILES: COC1OC2COC(OC2C(OC(=O)C2=CC=CC=C2)C1OC(=O)C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results